accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15793-77-8

IUPAC Name: 2-hydrazinylquinoline
Synonyms: 2-hydrazinylquinoline
Molecular Weight (g/mol): 159.19
Molecular Formula: C9H9N3
InChi Key: QMVCLSHKMIGEFN-UHFFFAOYSA-N
SMILES: NNC1=CC=C2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results