accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15801-69-1

IUPAC Name: 4-bromo-1,3,5-trimethyl-1H-pyrazole
Synonyms: 4-bromo-1,3,5-trimethylpyrazole
Molecular Weight (g/mol): 189.06
Molecular Formula: C6H9BrN2
InChi Key: UNTQXOJGXGRHMG-UHFFFAOYSA-N
SMILES: CN1N=C(C)C(Br)=C1C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results