CAS RN 1583-58-0
IUPAC Name:
2,4-difluorobenzoic acid
Synonyms:
2,4-difluorobenzoic acid
Molecular Weight (g/mol):
158.1
Molecular Formula:
C7H4F2O2
InChi Key:
NJYBIFYEWYWYAN-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=C(F)C=C1F
1
–
2
of
2
results
1
2,4-Difluorobenzoic acid, 98%
CAS: 1583-58-0 Molecular Formula: C7H4F2O2 Molecular Weight (g/mol): 158.10 MDL Number: MFCD00011670 InChI Key: NJYBIFYEWYWYAN-UHFFFAOYSA-N Synonym: 2,4-dfba,benzoic acid, 2,4-difluoro,2,4-difluorobenzoicacid,2,4-difluoro-benzoic acid,zlchem 472,pubchem1336,acmc-1brgf,2,4-difluorbenzoic acid,2,4-difluorobezoic acid,2 4-difluorobenzoic acid PubChem CID: 74102 IUPAC Name: 2,4-difluorobenzoic acid SMILES: OC(=O)C1=CC=C(F)C=C1F
2
2,4-Difluorobenzoic Acid, TRC
CAS: 1583-58-0 Molecular Formula: C7 H4 F2 O2 Molecular Weight (g/mol): 158.1 Synonym: 2,4-Difluorobenzoic acid,NSC 10312 IUPAC Name: 2,4-difluorobenzoic acid SMILES: OC(=O)c1ccc(F)cc1F