accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOBr

CAS RN 15848-22-3

IUPAC Name: 5-bromopentyl acetate
Synonyms: 5-bromopentyl acetate
Molecular Weight (g/mol): 209.08
Molecular Formula: C7H13BrO2
InChi Key: JIHJLQWUVZUKCH-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCCBr
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)