accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OONH2SH3C

CAS RN 15854-08-7

IUPAC Name: ethyl 2-amino-4-(4-methylphenyl)thiophene-3-carboxylate
Synonyms: ethyl 2-amino-4-(4-methylphenyl)thiophene-3-carboxylate
Molecular Weight (g/mol): 261.34
Molecular Formula: C14H15NO2S
InChi Key: GOQSMGNEJVWZIG-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC=C1C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results