accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OONH2SOH3C

CAS RN 15854-11-2

IUPAC Name: ethyl 2-amino-4-(4-methoxyphenyl)thiophene-3-carboxylate
Synonyms: ethyl 2-amino-4-(4-methoxyphenyl)thiophene-3-carboxylate
Molecular Weight (g/mol): 277.34
Molecular Formula: C14H15NO3S
InChi Key: ZUOUKGJUHCKVDK-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC=C1C1=CC=C(OC)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results