accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSNCS

CAS RN 15863-41-9

IUPAC Name: 1-isothiocyanato-4-(methylsulfanyl)benzene
Synonyms: 1-isothiocyanato-4-(methylsulfanyl)benzene
Molecular Weight (g/mol): 181.27
Molecular Formula: C8H7NS2
InChi Key: CEBAJHCAFXYWNT-UHFFFAOYSA-N
SMILES: CSC1=CC=C(C=C1)N=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)