accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOHOOH3COOH3C

CAS RN 1587-20-8

IUPAC Name: 1,2,3-trimethyl 2-hydroxypropane-1,2,3-tricarboxylate
Synonyms: 1,2,3-trimethyl 2-hydroxypropane-1,2,3-tricarboxylate
Molecular Weight (g/mol): 234.2
Molecular Formula: C9H14O7
InChi Key: HDDLVZWGOPWKFW-UHFFFAOYSA-N
SMILES: COC(=O)CC(O)(CC(=O)OC)C(=O)OC
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results