accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FSH

CAS RN 15894-04-9

IUPAC Name: (4-fluorophenyl)methanethiol
Synonyms: (4-fluorophenyl)methanethiol
Molecular Weight (g/mol): 142.19
Molecular Formula: C7H7FS
InChi Key: RKTRHMNWVZRZJQ-UHFFFAOYSA-N
SMILES: FC1=CC=C(CS)C=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results