accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3P

CAS RN 15912-75-1

IUPAC Name: triphenyl(propyl)phosphanium
Synonyms: triphenyl(propyl)phosphanium
Molecular Weight (g/mol): 305.38
Molecular Formula: C21H22P
InChi Key: MSXNDXIAUQJHNS-UHFFFAOYSA-N
SMILES: CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results