CAS RN 1592-20-7
IUPAC Name:
1-(chloromethyl)-4-ethenylbenzene
Synonyms:
4-vinylbenzyl chloride
Molecular Weight (g/mol):
152.62
Molecular Formula:
C9H9Cl
InChi Key:
ZRZHXNCATOYMJH-UHFFFAOYSA-N
SMILES:
ClCC1=CC=C(C=C)C=C1
1
–
2
of
2
results
1
Promotions Available
4-Vinylbenzyl chloride, 90%, technical
4-Vinylbenzyl chloride, 90%, technical
CAS: 1592-20-7 Molecular Formula: C9H9Cl Molecular Weight (g/mol): 152.62 MDL Number: MFCD00051362 InChI Key: ZRZHXNCATOYMJH-UHFFFAOYSA-N Synonym: 4-vinylbenzyl chloride,1-chloromethyl-4-vinylbenzene,4-chloromethylstyrene,4-vinylbenzylchloride,benzene, 1-chloromethyl-4-ethenyl,1-chloromethyl-4-ethenylbenzene,4-chloromethyl styrene,p-vinylbenzyl chloride,unii-cjl4oj6bs0,cjl4oj6bs0 PubChem CID: 74126 IUPAC Name: 1-(chloromethyl)-4-ethenylbenzene SMILES: ClCC1=CC=C(C=C)C=C1
2
4-(Chloromethyl)styrene (~0.1% TBC stabilizer), TRC
CAS: 1592-20-7 Molecular Formula: C9H9Cl Molecular Weight (g/mol): 152.62 IUPAC Name: 1-(chloromethyl)-4-ethenylbenzene SMILES: ClCc1ccc(C=C)cc1