accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1592-23-0

CaH3COOH3COO

CAS RN 1592-23-0

IUPAC Name: calcium dioctadecanoate
Synonyms: calcium stearate
Molecular Weight (g/mol): 607.03
Molecular Formula: C36H70CaO4
InChi Key: CJZGTCYPCWQAJB-UHFFFAOYSA-L
SMILES: [Ca++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (4)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)