accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOSNNOO

CAS RN 1594-56-5

IUPAC Name: [(2,4-dinitrophenyl)sulfanyl]formonitrile
Synonyms: DNTB 2,4-dinitrothiocyanatebenzene 2,4-dinitrothiocyanatobenzene 2,4-dinitrothiocyanobenzene 2,4-dinitro-rhodanbenzol
Molecular Weight (g/mol): 225.18
Molecular Formula: C7H3N3O4S
InChi Key: XQDQRCRASHAZBA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(SC#N)C(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results