accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1594-57-6

H2NNOHN(Z)

CAS RN 1594-57-6

IUPAC Name: 1-(1,4-dihydropyridin-4-ylidene)-1-nitrosomethanamine
Synonyms: 1-nitroso-1-(1H-pyridin-4-ylidene)methanamine
Molecular Weight (g/mol): 137.14
Molecular Formula: C6H7N3O
InChi Key: JEJGOULWKLAJFD-UHFFFAOYSA-N
SMILES: NC(N=O)=C1C=CNC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results