accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

RhOOOORhCCH3CCH3OOOOCCH3CH3C

CAS RN 15956-28-2

IUPAC Name: bis(λ²-rhodium(2+)) tetraacetate
Synonyms: rhodium(II) acetate
Molecular Weight (g/mol): 441.99
Molecular Formula: C8H12O8Rh2
InChi Key: SYBXSZMNKDOUCA-UHFFFAOYSA-J
SMILES: [Rh++].[Rh++].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)