accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClNHNCl

CAS RN 15965-30-7

IUPAC Name: 4,5-dichloro-1H-imidazole
Synonyms: 4,5-dichloro-1H-imidazole
Molecular Weight (g/mol): 136.96
Molecular Formula: C3H2Cl2N2
InChi Key: QAJJXHRQPLATMK-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)N=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results