accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NHClClN

CAS RN 15965-33-0

IUPAC Name: 4,5-dichloro-2-methyl-1H-imidazole
Synonyms: 4,5-dichloro-2-methyl-1H-imidazole
Molecular Weight (g/mol): 150.99
Molecular Formula: C4H4Cl2N2
InChi Key: ZWRYWVBALYOSPQ-UHFFFAOYSA-N
SMILES: CC1=NC(Cl)=C(Cl)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results