accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 159660-85-2

IUPAC Name: (3S)-1-benzyl-4-imino-3-(methoxycarbonyl)piperidin-1-ium
Synonyms: (3S)-1-benzyl-4-imino-3-(methoxycarbonyl)piperidin-1-ium
Molecular Weight (g/mol): 247.32
Molecular Formula: C14H19N2O2
InChi Key: XXLUTMHSBSWSBV-LBPRGKRZSA-O
SMILES: COC(=O)[C@H]1C[NH+](CC2=CC=CC=C2)CCC1=N
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results