accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 159709-36-1

IUPAC Name: 3,4-dimethylthieno[2,3-b]thiophene-2-carbaldehyde
Synonyms: 3,4-dimethylthieno[2,3-b]thiophene-2-carbaldehyde
Molecular Weight (g/mol): 196.28
Molecular Formula: C9H8OS2
InChi Key: WEAXSPMHCXWYPF-UHFFFAOYSA-N
SMILES: CC1=CSC2=C1C(C)=C(S2)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results