accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15980-15-1

IUPAC Name: 1,4-oxathiane
Synonyms: 1,4-oxathiane
Molecular Weight (g/mol): 104.17
Molecular Formula: C4H8OS
InChi Key: JBYHSSAVUBIJMK-UHFFFAOYSA-N
SMILES: C1CSCCO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results