accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16001-93-7

CH3OPOPOOH3COH3COH3C

CAS RN 16001-93-7

IUPAC Name: dimethyl [(dimethoxyphosphoryl)methyl]phosphonate
Synonyms: dimethyl (dimethoxyphosphoryl)methylphosphonate
Molecular Weight (g/mol): 232.11
Molecular Formula: C5H14O6P2
InChi Key: XAVFZUKFLWOSOS-UHFFFAOYSA-N
SMILES: COP(=O)(CP(=O)(OC)OC)OC
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)