accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CHNOOHNH2HCl

CAS RN 16012-55-8

IUPAC Name: 2-amino-3-(methylamino)propanoic acid hydrochloride
Synonyms: 3-(methylamino)-(DL)-alanine hydrochloride
Molecular Weight (g/mol): 154.59
Molecular Formula: C4H11ClN2O2
InChi Key: VDXYGASOGLSIDM-UHFFFAOYNA-N
SMILES: Cl.CNCC(N)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)