accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16018-83-0

ClH2NNH3NH2NNF(E/Z)(E/Z)

CAS RN 16018-83-0

IUPAC Name: N-[amino(azaniumyl)methylidene]-N''-(4-fluorophenyl)guanidine chloride
Synonyms: N-[amino(ammonio)methylidene]-N''-(4-fluorophenyl)guanidine chloride
Molecular Weight (g/mol): 231.66
Molecular Formula: C8H11ClFN5
InChi Key: HBYJTLZNKNUTCP-UHFFFAOYSA-N
SMILES: [Cl-].NC([NH3+])=NC(N)=NC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results