accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 160241-65-6

CH3NSHO

CAS RN 160241-65-6

IUPAC Name: 2-(2-methyl-1,3-thiazol-4-yl)phenol
Synonyms: 2-(2-methyl-1,3-thiazol-4-yl)phenol
Molecular Weight (g/mol): 191.25
Molecular Formula: C10H9NOS
InChi Key: LYLPLPWQCLBGHW-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C1=CC=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results