accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOI

CAS RN 160377-48-0

IUPAC Name: 5-(4-iodophenyl)-1,2-oxazole
Synonyms: 5-(4-iodophenyl)-1,2-oxazole
Molecular Weight (g/mol): 271.06
Molecular Formula: C9H6INO
InChi Key: TYNWYXFACGRXMG-UHFFFAOYSA-N
SMILES: IC1=CC=C(C=C1)C1=CC=NO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results