accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNOBr

CAS RN 160377-57-1

IUPAC Name: 3-(3-bromophenyl)-5-methyl-1,2,4-oxadiazole
Synonyms: 3-(3-bromophenyl)-5-methyl-1,2,4-oxadiazole
Molecular Weight (g/mol): 239.07
Molecular Formula: C9H7BrN2O
InChi Key: GTYLSVIVKRJHIQ-UHFFFAOYSA-N
SMILES: CC1=NC(=NO1)C1=CC(Br)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results