accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 160388-53-4

IUPAC Name: 4-[(1H-pyrazol-1-yl)methyl]benzoic acid
Synonyms: 4-(pyrazol-1-ylmethyl)benzoic acid
Molecular Weight (g/mol): 202.21
Molecular Formula: C11H10N2O2
InChi Key: ZHQQRHUITAFMTC-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(CN2C=CC=N2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results