accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNHOOH

CAS RN 16042-25-4

IUPAC Name: 1H-imidazole-2-carboxylic acid
Synonyms: 1H-imidazole-2-carboxylic acid
Molecular Weight (g/mol): 112.09
Molecular Formula: C4H4N2O2
InChi Key: KYWMCFOWDYFYLV-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results