accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 160446-22-0

CH3OOSOOCH3

CAS RN 160446-22-0

IUPAC Name: methyl 4-(methanesulfonylmethyl)benzoate
Synonyms: methyl 4-(methanesulfonylmethyl)benzoate
Molecular Weight (g/mol): 228.26
Molecular Formula: C10H12O4S
InChi Key: XCKJFFWCTKJUKD-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(CS(C)(=O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results