accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16063-69-7

IUPAC Name: 2,4,6-trichloropyridine
Synonyms: 2,4,6-trichloropyridine
Molecular Weight (g/mol): 182.43
Molecular Formula: C5H2Cl3N
InChi Key: FJNNGKMAGDPVIU-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=NC(Cl)=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results