accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16086-14-9

HOOHClCl

CAS RN 16086-14-9

IUPAC Name: 2,2-dichloroethane-1,1-diol
Synonyms: 2,2-dichloro-1,1-ethanediol C2H4cl2O2
Molecular Weight (g/mol): 130.95
Molecular Formula: C2H4Cl2O2
InChi Key: RUUBIFVWPACNLY-UHFFFAOYSA-N
SMILES: OC(O)C(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results