accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOOOCH3

CAS RN 1609-47-8

IUPAC Name: diethyl dicarbonate
Synonyms: diethyl pyrocarbonate ethyl pyrocarbonate oxydiformic acid diethyl ester ethoxyformic anhydride diethyl oxydiformate diethylpyrokarbonat diethylester kyseliny diuhlicite diethylpyrocarbonate dicarbonate, diethyl DKD
Molecular Weight (g/mol): 162.14
Molecular Formula: C6H10O5
InChi Key: FFYPMLJYZAEMQB-UHFFFAOYSA-N
SMILES: CCOC(=O)OC(=O)OCC
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (15)
  • (2)
Boiling Point 
  • (1)
  • (4)
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (5)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (15)
  • (2)
  • (1)
  • (4)
  • (1)
  • (5)
  • (2)
  • (8)
  • (9)
  • (5)
  • (2)