accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1609-86-5

CH3H3CCH3NCO

CAS RN 1609-86-5

IUPAC Name: 2-isocyanato-2-methylpropane
Synonyms: 2-isocyanato-2-methylpropane
Molecular Weight (g/mol): 99.13
Molecular Formula: C5H9NO
InChi Key: MGOLNIXAPIAKFM-UHFFFAOYSA-N
SMILES: CC(C)(C)N=C=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)