accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3HN

CAS RN 16096-32-5

IUPAC Name: 4-methyl-1H-indole
Synonyms: 4-methylindole
Molecular Weight (g/mol): 131.18
Molecular Formula: C9H9N
InChi Key: PZOUSPYUWWUPPK-UHFFFAOYSA-N
SMILES: CC1=C2C=CNC2=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results