accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3(R)NHONH2OHNOOFFF

CAS RN 160970-54-7

IUPAC Name: 1-(3-hydroxypropyl)-5-[(2R)-2-({2-[2-(2,2,2-trifluoroethoxy)phenoxy]ethyl}amino)propyl]-2,3-dihydro-1H-indole-7-carboxamide
Synonyms: silodosin
Molecular Weight (g/mol): 495.54
Molecular Formula: C25H32F3N3O4
InChi Key: PNCPYILNMDWPEY-QGZVFWFLSA-N
SMILES: C[C@H](CC1=CC2=C(N(CCCO)CC2)C(=C1)C(N)=O)NCCOC1=CC=CC=C1OCC(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)