accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHFBrCl

CAS RN 161045-79-0

IUPAC Name: 4-bromo-2-chloro-6-fluorophenol
Synonyms: 4-bromo-2-chloro-6-fluorophenol
Molecular Weight (g/mol): 225.44
Molecular Formula: C6H3BrClFO
InChi Key: QEYONPKSDTUPAX-UHFFFAOYSA-N
SMILES: OC1=C(F)C=C(Br)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results