accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3NHOH3CH3CCH3

CAS RN 1611-07-0

IUPAC Name: 2-(3,5-di-tert-butyl-4-hydroxyphenyl)acetonitrile
Synonyms: 2-(3,5-di-tert-butyl-4-hydroxyphenyl)acetonitrile
Molecular Weight (g/mol): 245.37
Molecular Formula: C16H23NO
InChi Key: BEQZSRGZHCDMMH-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(CC#N)=CC(=C1O)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results