accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 161290-85-3

IUPAC Name: 2-chloro-N-[4-(trifluoromethoxy)phenyl]acetamide
Synonyms: 2-chloro-N-[4-(trifluoromethoxy)phenyl]acetamide
Molecular Weight (g/mol): 253.61
Molecular Formula: C9H7ClF3NO2
InChi Key: MIWYZHHKNDZBCB-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(NC(=O)CCl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results