accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClHGeH2HCl

CAS RN 1613-66-7

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 301.77
Molecular Formula: C12H14Cl2Ge
InChi Key: YROQISGFJISKEN-UHFFFAOYSA-N
SMILES: Cl.Cl.[GeH2](C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)