accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 161321-36-4

OOHNHOO

CAS RN 161321-36-4

IUPAC Name: 2-cyclohexyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)acetic acid
Synonyms: cyclohexyl({[(9H-fluoren-9-ylmethoxy)carbonyl]amino})acetic acid
Molecular Weight (g/mol): 379.46
Molecular Formula: C23H25NO4
InChi Key: BWQQGHPODCJZDB-UHFFFAOYNA-N
SMILES: OC(=O)C(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)