accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OONNH2

CAS RN 16135-36-7

IUPAC Name: methyl 4-aminopyridine-3-carboxylate
Synonyms: methyl 4-aminopyridine-3-carboxylate
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: OCARFFAPQGYGBP-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results