accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 161395-97-7

BOOH3CCH3H3CH3COOH3C(E/Z)

CAS RN 161395-97-7

IUPAC Name: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)prop-2-en-1-yl acetate
Synonyms: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)prop-2-en-1-yl acetate
Molecular Weight (g/mol): 226.08
Molecular Formula: C11H19BO4
InChi Key: ZXBRLGZFCXGTBA-UHFFFAOYSA-N
SMILES: CC(=O)OCC=CB1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results