accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16143-84-3

CH3OHNFFFFFF

CAS RN 16143-84-3

IUPAC Name: N-[3,5-bis(trifluoromethyl)phenyl]acetamide
Synonyms: 3,5-diCF3 acetanilide
Molecular Weight (g/mol): 271.16
Molecular Formula: C10H7F6NO
InChi Key: XMKZELXFBZUGEY-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results