accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16156-50-6

H3COSOOH3C

CAS RN 16156-50-6

IUPAC Name: hexyl methanesulfonate
Synonyms: hexyl methanesulfonate
Molecular Weight (g/mol): 180.26
Molecular Formula: C7H16O3S
InChi Key: URIRDRHUUFRHAS-UHFFFAOYSA-N
SMILES: CCCCCCOS(C)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results