accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CPOOCH3OCH3

CAS RN 16165-68-7

IUPAC Name: diethyl decylphosphonate
Synonyms: diethyl decylphosphonate
Molecular Weight (g/mol): 278.37
Molecular Formula: C14H31O3P
InChi Key: HYQXIVAZQBYBAF-UHFFFAOYSA-N
SMILES: CCCCCCCCCCP(=O)(OCC)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results