accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 161660-94-2

CH3H3CCH3OOHN(S)(R)OO

CAS RN 161660-94-2

IUPAC Name: (1R,3S)-3-{[(tert-butoxy)carbonyl]amino}cyclopentane-1-carboxylate
Synonyms: (1R,3S)-3-[(tert-butoxycarbonyl)amino]cyclopentane-1-carboxylate
Molecular Weight (g/mol): 228.27
Molecular Formula: C11H18NO4
InChi Key: RNJQBGXOSAQQDG-SFYZADRCSA-M
SMILES: CC(C)(C)OC(=O)N[C@H]1CC[C@H](C1)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)