accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16179-97-8

IUPAC Name: 2-(pyridin-2-yl)acetic acid hydrochloridyl
Synonyms: pyridin-2-ylacetic acid hydrochloridyl
Molecular Weight (g/mol): 172.59
Molecular Formula: C7H7ClNO2
InChi Key: MRNWGZIWHQZCDS-UHFFFAOYSA-N
SMILES: [Cl].OC(=O)CC1=CC=CC=N1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)