accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1618-26-4

IUPAC Name: bis(methylsulfanyl)methane
Synonyms: 2,4-dithiapentane
Molecular Weight (g/mol): 108.22
Molecular Formula: C3H8S2
InChi Key: LOCDPORVFVOGCR-UHFFFAOYSA-N
SMILES: CSCSC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results