accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3H3CSOOHNNH2

CAS RN 16182-15-3

IUPAC Name: 2,4,6-trimethylbenzene-1-sulfonohydrazide
Synonyms: 2,4,6-trimethylbenzenesulfonohydrazide
Molecular Weight (g/mol): 214.28
Molecular Formula: C9H14N2O2S
InChi Key: JQUBKTQDNVZHIY-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C(C)=C1)S(=O)(=O)NN
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)