accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1619994-68-1

IUPAC Name: 1-[1-(2-methanesulfonylphenyl)-7-propoxyindolizin-3-yl]ethan-1-one
Synonyms: 1-[1-(2-methanesulfonylphenyl)-7-propoxyindolizin-3-yl]ethanone
Molecular Weight (g/mol): 371.45
Molecular Formula: C20H21NO4S
InChi Key: KHWCPNJRJCNVRI-UHFFFAOYSA-N
SMILES: CCCOC1=CC2=C(C=C(N2C=C1)C(C)=O)C1=CC=CC=C1S(C)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results